Potassium vinyltrifluoroborate, 97%
Potassium vinyltrifluoroborate, 97%
Potassium vinyltrifluoroborate, 97%
Thermo Scientific Chemicals

Potassium vinyltrifluoroborate, 97%

CAS: 13682-77-4 | C2H3BF3K | 133.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L17970.06
also known as L17970-06
Price (USD)
154.65
Special offer
Online exclusive
Ends: 30-Jun-2026
182.00
Save 27.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
154.65
Special offer
Online exclusive
Ends: 30-Jun-2026
182.00
Save 27.35 (15%)
Each
Chemical Identifiers
CAS13682-77-4
IUPAC Namepotassium ethenyltrifluoroboranuide
Molecular FormulaC2H3BF3K
InChI KeyZCUMGICZWDOJEM-UHFFFAOYSA-N
SMILES[K+].F[B-](F)(F)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)≤1.0%
Proton NMRConforms to structure
Appearance (Color)White to pale cream
FormPowder or crystalline powder or lumps
Assay≥96.0%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only