Potassium trans-beta-styryltrifluoroborate, 98%
Potassium trans-beta-styryltrifluoroborate, 98%
Potassium trans-beta-styryltrifluoroborate, 98%
Potassium trans-beta-styryltrifluoroborate, 98%
Thermo Scientific Chemicals

Potassium trans-beta-styryltrifluoroborate, 98%

CAS: 201852-49-5 | C8H7BF3K | 210.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L17971.03
also known as L17971-03
Price (USD)
56.65
Special offer
Online exclusive
Ends: 30-Jun-2026
65.90
Save 9.25 (14%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
56.65
Special offer
Online exclusive
Ends: 30-Jun-2026
65.90
Save 9.25 (14%)
Each
Add to cart
Chemical Identifiers
CAS201852-49-5
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Elemental AnalysisB: 5.04-5.25% (Theory 5.14%)
Elemental AnalysisH: 3.29-3.43% (Theory 3.35%)
Elemental AnalysisK: 18.24-18.98% (Theory 18.61%)
Appearance (Color)White
View more
Potassium trans-beta-styryltrifluoroborate is used as a reactant for Suzuki-Miyaura cross-coupling, in enantioselective total synthesis of naseseazines A and B via regioselective Friedel-Crafts based arylative dimerization, in metal-free chlorodeboronation reactions, in preparation of aryl ketones via Lewis acid-catalyzed nucleophilic substitution and in preparation of Weinreb amides by palladium-catalyzed cross-coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium trans-beta-styryltrifluoroborate is used as a reactant for Suzuki-Miyaura cross-coupling, in enantioselective total synthesis of naseseazines A and B via regioselective Friedel-Crafts based arylative dimerization, in metal-free chlorodeboronation reactions, in preparation of aryl ketones via Lewis acid-catalyzed nucleophilic substitution and in preparation of Weinreb amides by palladium-catalyzed cross-coupling reactions.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. El Bakali, J., Maingot, L., Dumont, J., Host, H., Hocine, A., Cousaert, N., ... & Deprez-Poulain, R. (2012). Novel selective inhibitors of neutral endopeptidase: discovery by screening and hit-to-lead optimisation. MedChemComm, 3(4), 469-474.