N,N'-Dibenzylethylenediamine diacetate, 99%
N,N'-Dibenzylethylenediamine diacetate, 99%
N,N'-Dibenzylethylenediamine diacetate, 99%
Thermo Scientific Chemicals

N,N'-Dibenzylethylenediamine diacetate, 99%

CAS: 122-75-8 | C20H28N2O4 | 360.454 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number L17974.14
also known as L17974-14
Price (USD)
29.65
Online Exclusive
32.80
Save 3.15 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
29.65
Online Exclusive
32.80
Save 3.15 (10%)
Each
Chemical Identifiers
CAS122-75-8
IUPAC Namebis(acetic acid); benzyl[2-(benzylamino)ethyl]amine
Molecular FormulaC20H28N2O4
InChI KeyMTRNNCLQPVCDLF-UHFFFAOYSA-N
SMILESCC(O)=O.CC(O)=O.C(CNCC1=CC=CC=C1)NCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Melting Point (clear melt)113.0-119.0°C
Appearance (Color)White to pale cream
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only