1-Boc-indole-2-boronic acid, 95%
1-Boc-indole-2-boronic acid, 95%
1-Boc-indole-2-boronic acid, 95%
1-Boc-indole-2-boronic acid, 95%
Thermo Scientific Chemicals

1-Boc-indole-2-boronic acid, 95%

CAS: 213318-44-6 | C13H16BNO4 | 261.084 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number L18009.03
also known as L18009-03
Price (USD)
95.65
Special offer
Online exclusive
Ends: 30-Jun-2026
112.00
Save 16.35 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
95.65
Special offer
Online exclusive
Ends: 30-Jun-2026
112.00
Save 16.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS213318-44-6
IUPAC Name{1-[(tert-butoxy)carbonyl]-1H-indol-2-yl}boronic acid
Molecular FormulaC13H16BNO4
InChI KeySVIBPSNFXYUOFT-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)N1C(=CC2=CC=CC=C12)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown or pink
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥94.0%
Proton NMRConforms to structure
Assay (HPLC)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only