Bis(tetramethylene)fluoroformamidinium hexafluorophosphate
Bis(tetramethylene)fluoroformamidinium hexafluorophosphate
Bis(tetramethylene)fluoroformamidinium hexafluorophosphate
Thermo Scientific Chemicals

Bis(tetramethylene)fluoroformamidinium hexafluorophosphate

CAS: 164298-25-3 | C9H16F7N2P | 316.204 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L18013.03
also known as L18013-03
Price (USD)
56.65
Online Exclusive
62.80
Save 6.15 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
56.65
Online Exclusive
62.80
Save 6.15 (10%)
Each
Chemical Identifiers
CAS164298-25-3
IUPAC Name1-[fluoro(pyrrolidin-1-yl)methylidene]-1λ⁵-pyrrolidin-1-ylium; hexafluoro-λ⁵-phosphanuide
Molecular FormulaC9H16F7N2P
InChI KeyMNJUGQKOHJQOCK-UHFFFAOYSA-N
SMILESF[P-](F)(F)(F)(F)F.FC(N1CCCC1)=[N+]1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Elemental Analysis(Carbon, C) : 33.1-35.2%%
FormPowder
Elemental Analysis(Nitrogen, N) : 8.59-9.12%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only