3-(Bromomethyl)phenoxyacetic acid, 97%
3-(Bromomethyl)phenoxyacetic acid, 97%
3-(Bromomethyl)phenoxyacetic acid, 97%
3-(Bromomethyl)phenoxyacetic acid, 97%
Thermo Scientific Chemicals

3-(Bromomethyl)phenoxyacetic acid, 97%

CAS: 136645-25-5 | C9H9BrO3 | 245.072 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L18029.03
also known as L18029-03
Price (USD)
39.65
Special offer
Online exclusive
Ends: 30-Jun-2026
46.10
Save 6.45 (14%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
39.65
Special offer
Online exclusive
Ends: 30-Jun-2026
46.10
Save 6.45 (14%)
Each
Add to cart
Chemical Identifiers
CAS136645-25-5
IUPAC Name2-[3-(bromomethyl)phenoxy]acetic acid
Molecular FormulaC9H9BrO3
InChI KeyMSTODKGDFXWAIO-UHFFFAOYSA-N
SMILESOC(=O)COC1=CC=CC(CBr)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (HPLC)≥96.0%
Melting Point (clear melt)122.5-128.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only