3-(4-Methoxybenzoyl)acrylic acid, 98%
3-(4-Methoxybenzoyl)acrylic acid, 98%
3-(4-Methoxybenzoyl)acrylic acid, 98%
Thermo Scientific Chemicals

3-(4-Methoxybenzoyl)acrylic acid, 98%

CAS: 5711-41-1 | C11H10O4 | 206.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L18071.03
also known as L18071-03
Price (USD)
47.65
Special offer
Online exclusive
Ends: 28-Dec-2026
55.70
Save 8.05
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
47.65
Special offer
Online exclusive
Ends: 28-Dec-2026
55.70
Save 8.05
Each
Chemical Identifiers
CAS5711-41-1
IUPAC Name(2E)-4-(4-methoxyphenyl)-4-oxobut-2-enoic acid
Molecular FormulaC11H10O4
InChI KeyWORYXBDHTBWLLL-VOTSOKGWSA-N
SMILESCOC1=CC=C(C=C1)C(=O)C=CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA≥97.5% (U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
Appearance (Color)Yellow to yellow-grey
FormPowder
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5% (non-U.S. specification)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only