1,6-Dimethoxynaphthalene, 98+%
1,6-Dimethoxynaphthalene, 98+%
1,6-Dimethoxynaphthalene, 98+%
Thermo Scientific Chemicals

1,6-Dimethoxynaphthalene, 98+%

CAS: 3900-49-0 | C12H12O2 | 188.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L18129.06
also known as L18129-06
Price (USD)
180.65
Online Exclusive
201.00
Save 20.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
180.65
Online Exclusive
201.00
Save 20.35 (10%)
Each
Chemical Identifiers
CAS3900-49-0
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Melting Point (clear melt)54.0-61.0?C
FormCrystals or powder or crystalline powder
Appearance (Color)White to pale yellow to yellow to brown to gray
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only