2,4-Bis(trifluoromethyl)benzeneboronic acid, 97%
2,4-Bis(trifluoromethyl)benzeneboronic acid, 97%
2,4-Bis(trifluoromethyl)benzeneboronic acid, 97%
Thermo Scientific Chemicals

2,4-Bis(trifluoromethyl)benzeneboronic acid, 97%

CAS: 153254-09-2 | C8H5BF6O2 | 257.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L18161.06
also known as L18161-06
Price (USD)
194.65
Special offer
Online exclusive
Ends: 30-Jun-2026
229.00
Save 34.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
194.65
Special offer
Online exclusive
Ends: 30-Jun-2026
229.00
Save 34.35 (15%)
Each
Chemical Identifiers
CAS153254-09-2
IUPAC Name[2,4-bis(trifluoromethyl)phenyl]boronic acid
Molecular FormulaC8H5BF6O2
InChI KeyWLYPBMBWKYALCG-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(C=C1C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥96.0%
Appearance (Color)White to cream
Melting Point (clear melt)141.0-150.0?C
FormPowder
Assay (HPLC)≥96.0%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only