1-Methyl-1,2,4-triazole, 98%
1-Methyl-1,2,4-triazole, 98%
1-Methyl-1,2,4-triazole, 98%
Thermo Scientific Chemicals

1-Methyl-1,2,4-triazole, 98%

CAS: 6086-21-1 | C3H5N3 | 83.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L18337.03
also known as L18337-03
Price (USD)
51.65
Special offer
Online exclusive
Ends: 30-Jun-2026
60.90
Save 9.25 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
51.65
Special offer
Online exclusive
Ends: 30-Jun-2026
60.90
Save 9.25 (15%)
Each
Chemical Identifiers
CAS6086-21-1
IUPAC Name1-methyl-1H-1,2,4-triazole
Molecular FormulaC3H5N3
InChI KeyMWZDIEIXRBWPLG-UHFFFAOYSA-N
SMILESCN1C=NC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
Refractive Index1.4685-1.4725 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only