3,4,5-Trifluorobenzeneboronic acid, 97%
3,4,5-Trifluorobenzeneboronic acid, 97%
3,4,5-Trifluorobenzeneboronic acid, 97%
Thermo Scientific Chemicals

3,4,5-Trifluorobenzeneboronic acid, 97%

CAS: 143418-49-9 | C6H4BF3O2 | 175.901 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L18519.03
also known as L18519-03
Price (USD)
166.65
Special offer
Online exclusive
Ends: 28-Dec-2026
196.00
Save 29.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
166.65
Special offer
Online exclusive
Ends: 28-Dec-2026
196.00
Save 29.35
Each
Chemical Identifiers
CAS143418-49-9
IUPAC Name(3,4,5-trifluorophenyl)boronic acid
Molecular FormulaC6H4BF3O2
InChI KeyUHDDEIOYXFXNNJ-UHFFFAOYSA-N
SMILESOB(O)C1=CC(F)=C(F)C(F)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale brown
Assay from Supplier's CofA≥96.0% (HPLC)
FormPowder
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only