1-Octylphosphonic acid, 99%
1-Octylphosphonic acid, 99%
1-Octylphosphonic acid, 99%
Thermo Scientific Chemicals

1-Octylphosphonic acid, 99%

CAS: 4724-48-5 | C8H19O3P | 194.211 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L18586.06
also known as L18586-06
Price (USD)
269.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
269.00
Each
Chemical Identifiers
CAS4724-48-5
IUPAC Nameoctylphosphonic acid
Molecular FormulaC8H19O3P
InChI KeyNJGCRMAPOWGWMW-UHFFFAOYSA-N
SMILESCCCCCCCCP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)96.0-102.0?C
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
1-Octylphosphonic acid is a basic building block and used in the preparation of various pharmaceutical compounds and chemical products. It is supported with calcium hydroxyapatite and used in the dental field as corrosion inhibitor and organic building block. It acts as a corrosion inhibitor in metallic household surfaces such as faucets and drains as well as prevents the rusting of can or dispenser. It is also used in the synthesis of octyl-phosphonic acid dimethyl ester by reacting with iodomethane in presence of CS2 CO3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octylphosphonic acid is a basic building block and used in the preparation of various pharmaceutical compounds and chemical products. It is supported with calcium hydroxyapatite and used in the dental field as corrosion inhibitor and organic building block. It acts as a corrosion inhibitor in metallic household surfaces such as faucets and drains as well as prevents the rusting of can or dispenser. It is also used in the synthesis of octyl-phosphonic acid dimethyl ester by reacting with iodomethane in presence of CS2 CO3.

Solubility
Slightly soluble in organic solvents and water.

Notes
Incompatible with strong oxidizing agents. Keep the container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. Wang, H.; Mativetsky, J. M.; Ren, Y.; Gomez, E. D.; Jaye, C.; Schwartz, J.; Fischer, D. A.; Loo, Y. L. Fluorinated and hydrogenated self-assembled monolayers (SAMs) on anodes: Effects of SAM chemistry on device characteristics of polymer solar cells. Org. Electron. 2014, 15 (11) 3333-3340.
  2. Debrassi, A.; Ribbera, a.; Vos, W. M.; Wennekes, T.; Zuilhof, H. Stability of (Bio)Functionalized Porous Aluminum Oxide. Langmuir 2014, 30 (5), 1311-1320.