1-Octylphosphonic dichloride is employed in the surface modification of calcium fluoro and hydroxyapatite by 1-octylphosphonic dichloride and qualitative and quantitative analysis of phosphate rock from Hazm Al-jalamid area, northern Saudi Arabia.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Octylphosphonic dichloride is employed in the surface modification of calcium fluoro and hydroxyapatite by 1-octylphosphonic dichloride and qualitative and quantitative analysis of phosphate rock from Hazm Al-jalamid area, northern Saudi Arabia.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from strong bases, water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only
General References:
- Abdallah Aissa, et al. Surface modification of calcium fluoro and hydroxyapatite by 1-octylphosphonic dichloride.Appl. Surf. Sci.,2011,257(21), 9002-9007.
- Abdallah Aissa, et al. Qualitative and quantitative analysis of phosphate rock from Hazm Al-jalamid area, northern Saudi Arabia.International Journal of Basic and Applied Sciences.,2014,3(3), 190-198.