1,2,4-Triazolo[4,3-a]pyridin-3(2H)-one, 98+%
1,2,4-Triazolo[4,3-a]pyridin-3(2H)-one, 98+%
1,2,4-Triazolo[4,3-a]pyridin-3(2H)-one, 98+%
Thermo Scientific Chemicals

1,2,4-Triazolo[4,3-a]pyridin-3(2H)-one, 98+%

CAS: 6969-71-7 | C6H5N3O | 135.126 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L18705.14
also known as L18705-14
Price (USD)
77.65
Online Exclusive
85.70
Save 8.05 (9%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
77.65
Online Exclusive
85.70
Save 8.05 (9%)
Each
Chemical Identifiers
CAS6969-71-7
IUPAC Name2H,3H-[1,2,4]triazolo[4,3-a]pyridin-3-one
Molecular FormulaC6H5N3O
InChI KeyLJRXNXBFJXXRNQ-UHFFFAOYSA-N
SMILESO=C1NN=C2C=CC=CN12
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)231.5-237.5?C
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (HPLC)≥98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only