1,1,3,3-Tetramethylbutyl isocyanate, 98%
1,1,3,3-Tetramethylbutyl isocyanate, 98%
1,1,3,3-Tetramethylbutyl isocyanate, 98%
Thermo Scientific Chemicals

1,1,3,3-Tetramethylbutyl isocyanate, 98%

CAS: 1611-57-0 | C9H17NO | 155.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L18706.06
also known as L18706-06
Price (USD)
65.65
Special offer
Online exclusive
Ends: 30-Jun-2026
77.20
Save 11.55 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
65.65
Special offer
Online exclusive
Ends: 30-Jun-2026
77.20
Save 11.55 (15%)
Each
Chemical Identifiers
CAS1611-57-0
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4260-1.4300 @ 20?C
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only