(S)-(+)-6-Methyl-5-hepten-2-ol, 99%
(S)-(+)-6-Methyl-5-hepten-2-ol, 99%
(S)-(+)-6-Methyl-5-hepten-2-ol, 99%
(S)-(+)-6-Methyl-5-hepten-2-ol, 99%
Thermo Scientific Chemicals

(S)-(+)-6-Methyl-5-hepten-2-ol, 99%

CAS: 58917-26-3 | C8H16O | 128.215 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L18884.MD
also known as L18884-MD
Price (USD)
53.65
Special offer
Online exclusive
Ends: 30-Jun-2026
62.70
Save 9.05 (14%)
Each
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)
53.65
Special offer
Online exclusive
Ends: 30-Jun-2026
62.70
Save 9.05 (14%)
Each
Add to cart
Chemical Identifiers
CAS58917-26-3
IUPAC Name6-methylhept-5-en-2-ol
Molecular FormulaC8H16O
InChI KeyOHEFFKYYKJVVOX-UHFFFAOYNA-N
SMILESCC(O)CCC=C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Rotation18.1 ± 0.1? (neat)
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
Refractive Index1.4470-1.4500 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only