2-(2-Aminoethoxy)ethanol, 98%
2-(2-Aminoethoxy)ethanol, 98%
2-(2-Aminoethoxy)ethanol, 98%
2-(2-Aminoethoxy)ethanol, 98%
Thermo Scientific Chemicals

2-(2-Aminoethoxy)ethanol, 98%

CAS: 929-06-6 | C4H11NO2 | 105.137 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number L18897.22
also known as L18897-22
Price (USD)
39.65
Special offer
Online exclusive
Ends: 30-Jun-2026
46.30
Save 6.65 (14%)
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
39.65
Special offer
Online exclusive
Ends: 30-Jun-2026
46.30
Save 6.65 (14%)
Each
Add to cart
Chemical Identifiers
CAS929-06-6
IUPAC Name2-(2-aminoethoxy)ethan-1-ol
Molecular FormulaC4H11NO2
InChI KeyGIAFURWZWWWBQT-UHFFFAOYSA-N
SMILESNCCOCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)≥97.5%
Refractive Index1.4570-1.4610 @ 20?C
FormLiquid
2-(2-Aminoethoxy)ethanol is used mainly in stripper solutions for applications in electronics, in gas treating to remove carbon dioxide and hydrogen sulfide, and in coolants/lubricants for metal working applications. Other applications of aminoethoxyethanol (ADG) include, inter alia, crop protection products, surfactants and colorants. It is also used as a selective solvent for recovery of aromatics from refinery streams and in preparation of foam stabilizers, wetting and emulsifying agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Aminoethoxy)ethanol is used mainly in stripper solutions for applications in electronics, in gas treating to remove carbon dioxide and hydrogen sulfide, and in coolants/lubricants for metal working applications. Other applications of aminoethoxyethanol (ADG) include, inter alia, crop protection products, surfactants and colorants. It is also used as a selective solvent for recovery of aromatics from refinery streams and in preparation of foam stabilizers, wetting and emulsifying agents.

Solubility
Fully miscible in water, alcohols and aromatic hydrocarbons.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive to air. Store under inert gas.
RUO – Research Use Only

General References:

  1. Rob J. Littel, Martinus Bos, Gerdine J. Knoop. Dissociation constants of some alkanolamines at 293, 303, 318, and 333 K. J. Chem. Eng. Data. 1990, 35 (3), 276-277.
  2. Baris Kiskan.; Yusuf Yagc.; Hatsuo Ishida. Synthesis, characterization, and properties of new thermally curable polyetheresters containing benzoxazine moieties in the main chain. Journal of Polymer Science Part A: Polymer Chemistry. 2008, 46 (2), 414-420.