Methyl nitroacetate, 97+%
Methyl nitroacetate, 97+%
Methyl nitroacetate, 97+%
Thermo Scientific Chemicals

Methyl nitroacetate, 97+%

CAS: 2483-57-0 | C3H5NO4 | 119.076 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number L18950.09
also known as L18950-09
Price (USD)
282.65
Online Exclusive
314.00
Save 31.35 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
282.65
Online Exclusive
314.00
Save 31.35 (10%)
Each
Chemical Identifiers
CAS2483-57-0
IUPAC Namemethyl 2-nitroacetate
Molecular FormulaC3H5NO4
InChI KeyALBSWLMUHHZLLR-UHFFFAOYSA-N
SMILESCOC(=O)C[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA≥96.0% (U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
Identification (FTIR)Conforms (non-U.S. specification)
Refractive Index1.4235-1.4285 @ 20°C (non-U.S. specification)
Appearance (Color)Clear colorless to yellow
View more
Methyl nitroacetate is used in the synthesis of aromatic and heteroaromatic linear (E)-alfa-nitro-arylpentenoates. It is also used to prepare phenyliodonium ylide. Further, it is utilized in the highly enantioselective and diastereoselective copper(I)-catalyzed cyclopropanation of alkenes. In addition to this, it is employed in the production of methyl (Z)-2-nitro-3-(4-nitrophenyl)-2-propenoate through reaction with 4-nitrobenzylideneaniline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl nitroacetate is used in the synthesis of aromatic and heteroaromatic linear (E)-alfa-nitro-arylpentenoates. It is also used to prepare phenyliodonium ylide. Further, it is utilized in the highly enantioselective and diastereoselective copper(I)-catalyzed cyclopropanation of alkenes. In addition to this, it is employed in the production of methyl (Z)-2-nitro-3-(4-nitrophenyl)-2-propenoate through reaction with 4-nitrobenzylideneaniline.

Solubility
Miscible with ethanol, ether and most organic solvents.

Notes
Light sensitive. Incompatible withstrong bases.
RUO – Research Use Only

General References:

  1. Hoffmann, R. C.; Schneider, J. J. Synergistic and Antagonistic Ligand Effects in the Transformation of Silver Compounds of Keto- and Cyano-Functionalised Oximates and Nitronates: A Systematic Study Using Thermal Analysis. Eur. J. Inorg. Chem. 2015, 2015 (25), 4254-4259.
  2. Afreh, I.; Wujcik, E. K.; Blasdel, N.; Sauer, B.; Kaya, S.; Duirk, S.; Monty, C. N. Detection of halogenated organics by their inhibitory action in a catalytic reaction between dimethyl acetylenedicarboxylate and 2-methyl-4-nitroaniline. J. Anal. Chem. 2015, 70 (7), 825-830.