(S)-(-)-3-Hydroxy-3-phenylpropionic acid, 98+%
(S)-(-)-3-Hydroxy-3-phenylpropionic acid, 98+%
(S)-(-)-3-Hydroxy-3-phenylpropionic acid, 98+%
Thermo Scientific Chemicals

(S)-(-)-3-Hydroxy-3-phenylpropionic acid, 98+%

CAS: 36567-72-3 | C9H10O3 | 166.176 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L19041.MD
also known as L19041-MD
Price (USD)
62.65
Special offer
Online exclusive
Ends: 30-Jun-2026
73.70
Save 11.05 (15%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
62.65
Special offer
Online exclusive
Ends: 30-Jun-2026
73.70
Save 11.05 (15%)
Each
Chemical Identifiers
CAS36567-72-3
IUPAC Name(3S)-3-hydroxy-3-phenylpropanoate
Molecular FormulaC9H9O3
InChI KeyAYOLELPCNDVZKZ-QMMMGPOBSA-M
SMILESO[C@@H](CC([O-])=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Optical Rotation-22.6 ± 0.5? (C=4 in methanol)
Melting Point (clear melt)115.0-122.0?C
FormCrystals or powder or crystalline powder or fused solid
Assay (Silylated GC)≥98.0%
Appearance (Color)White to pale cream
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only