2-Methylindole-3-acetic acid, 98+%
2-Methylindole-3-acetic acid, 98+%
2-Methylindole-3-acetic acid, 98+%
Thermo Scientific Chemicals

2-Methylindole-3-acetic acid, 98+%

CAS: 1912-43-2 | C11H11NO2 | 189.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L19071.06
also known as L19071-06
Price (USD)
94.65
Online Exclusive
105.00
Save 10.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
94.65
Online Exclusive
105.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS1912-43-2
IUPAC Name2-(2-methyl-1H-indol-3-yl)acetic acid
Molecular FormulaC11H11NO2
InChI KeyQJNNHJVSQUUHHE-UHFFFAOYSA-N
SMILESCC1=C(CC(O)=O)C2=CC=CC=C2N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (HPLC)≥98.0%
Appearance (Color)White to cream or pale pink
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only