5-Methoxyindole-3-carboxaldehyde, 99%
5-Methoxyindole-3-carboxaldehyde, 99%
5-Methoxyindole-3-carboxaldehyde, 99%
Thermo Scientific Chemicals

5-Methoxyindole-3-carboxaldehyde, 99%

CAS: 10601-19-1 | C10H9NO2 | 175.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L19072.06
also known as L19072-06
Price (USD)
279.65
Online Exclusive
310.00
Save 30.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
279.65
Online Exclusive
310.00
Save 30.35 (10%)
Each
Chemical Identifiers
CAS10601-19-1
IUPAC Name5-methoxy-1H-indole-3-carbaldehyde
Molecular FormulaC10H9NO2
InChI KeyTUWARWGEOHQXCO-UHFFFAOYSA-N
SMILESCOC1=CC=C2NC=C(C=O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.5%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)179.0-185.0?C
Appearance (Color)Yellow
Free acid (titration)≤0.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only