Benzenesulfonic acid, tech. ca 75% w/w aq. soln.
Benzenesulfonic acid, tech. ca 75% w/w aq. soln.
Benzenesulfonic acid, tech. ca 75% w/w aq. soln.
Thermo Scientific Chemicals

Benzenesulfonic acid, tech. ca 75% w/w aq. soln.

CAS: 98-11-3 | C6H6O3S | 158.171 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1000 mL
5000 mL
Catalog number L19077.0J
also known as L19077-0J
Price (USD)
119.65
Special offer
Online exclusive
Ends: 30-Jun-2026
141.00
Save 21.35 (15%)
Each
Quantity:
5000 mL
Request bulk or custom format
Price (USD)
119.65
Special offer
Online exclusive
Ends: 30-Jun-2026
141.00
Save 21.35 (15%)
Each
Chemical Identifiers
CAS98-11-3
IUPAC Namebenzenesulfonic acid
Molecular FormulaC6H6O3S
InChI KeySRSXLGNVWSONIS-UHFFFAOYSA-N
SMILESOS(=O)(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colourless to dark brown
Assay (Aqueous acid-base Titration)> 73.0%
Formliquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only