Thermo Scientific Chemicals

4-Bromophenylacetylene, 97%, Thermo Scientific Chemicals

Catalog number: L19082.MD
250 mg, Each
Thermo Scientific Chemicals

4-Bromophenylacetylene, 97%, Thermo Scientific Chemicals

Catalog number: L19082.MD
250 mg, Each
Quantity
Catalog number: L19082.MD
also known as L19082-MD
Price (USD)
Quantity
-

Chemical Identifiers

CAS
766-96-1
IUPAC Name
1-bromo-4-ethynylbenzene
Molecular Formula
C8H5Br
InChI Key
LTLVZQZDXQWLHU-UHFFFAOYSA-N
SMILES
BrC1=CC=C(C=C1)C#C
Identification (FTIR)
Conforms
Melting Point (clear melt)
59.0-68.0°C
Appearance (Color)
White to pale cream or pale yellow
Form
Crystals or powder or crystalline powder
Assay (GC)
≥96.0%

Description

4-Bromophenylacetylene, is used as the starting material for second-order nonlinear optical materials, heterocyclotriynes, and unsymmetrical 1,4-diarylbutadiynes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromophenylacetylene, is used as the starting material for second-order nonlinear optical materials, heterocyclotriynes, and unsymmetrical 1,4-diarylbutadiynes.

Solubility
Soluble in chloroform, diethyl ether and ethanol.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text