(R)-(-)-Mandelic acid, ChiPros 99+%, ee 99+%
(R)-(-)-Mandelic acid, ChiPros 99+%, ee 99+%
(R)-(-)-Mandelic acid, ChiPros 99+%, ee 99+%
Thermo Scientific Chemicals

(R)-(-)-Mandelic acid, ChiPros 99+%, ee 99+%

CAS: 611-71-2 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L19116.14
also known as L19116-14
Price (USD)
78.65
Online Exclusive
87.70
Save 9.05 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
78.65
Online Exclusive
87.70
Save 9.05 (10%)
Each
Chemical Identifiers
CAS611-71-2
IUPAC Name(2R)-2-hydroxy-2-phenylacetic acid
Molecular FormulaC8H8O3
InChI KeyIWYDHOAUDWTVEP-SSDOTTSWSA-N
SMILESO[C@@H](C(O)=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystalline powder
Assay (Aqueous acid-base Titration)>99.0 to <101.00%
Enantiomeric excess>99% by HPLC
Optical Rotation-154? ± 4? (c=5, water)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only