(R)-(+)-1-(4-Chlorophenyl)ethylamine, ChiPros™ 97%, ee 98%
(R)-(+)-1-(4-Chlorophenyl)ethylamine, ChiPros™ 97%, ee 98%
(R)-(+)-1-(4-Chlorophenyl)ethylamine, ChiPros™ 97%, ee 98%
(R)-(+)-1-(4-Chlorophenyl)ethylamine, ChiPros™ 97%, ee 98%
Thermo Scientific Chemicals

(R)-(+)-1-(4-Chlorophenyl)ethylamine, ChiPros™ 97%, ee 98%

CAS: 27298-99-3 | C8H10ClN | 155.625 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L19119.06
also known as L19119-06
Price (USD)
109.65
Online Exclusive
122.00
Save 12.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
109.65
Online Exclusive
122.00
Save 12.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS27298-99-3
IUPAC Name1-(4-chlorophenyl)ethan-1-amine
Molecular FormulaC8H10ClN
InChI KeyPINPOEWMCLFRRB-UHFFFAOYNA-N
SMILESCC(N)C1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.5400-1.5440 @ 20?C
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (GC)>96.0%
Optical Rotation+31? ± 2? (neat)
View more
(R)-(+)-1-(4-Chlorophenyl)ethylamine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as a chiral reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-1-(4-Chlorophenyl)ethylamine, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is used as a chiral reagent.

Solubility
Soluble in DMSO, insoluble in water.

Notes
Air Sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

General References:

  1. DT Guranda.; LM van Langen.; F van Rantwijk. Highly efficient and enantioselective enzymatic acylation of amines in aqueous medium. Tetrahedron: Asymmetry. 2001 12 (11), 1645-1650.
  2. Y Itabashi.; A Kukis.; L Marai.; T Takagi. HPLC resolution of diacylglycerol moieties of natural triacylglycerols on a chiral phase consisting of bonded (R)-(+)-1-(1-naphthyl) ethylamine. Journal of lipid research. 1990 31 (1), 1711-1717.