(S)-(-)-1-(4-Fluorophenyl)ethylamine, ChiPros 99%, ee 99%
(S)-(-)-1-(4-Fluorophenyl)ethylamine, ChiPros 99%, ee 99%
(S)-(-)-1-(4-Fluorophenyl)ethylamine, ChiPros 99%, ee 99%
Thermo Scientific Chemicals

(S)-(-)-1-(4-Fluorophenyl)ethylamine, ChiPros 99%, ee 99%

CAS: 66399-30-2 | C8H10FN | 139.173 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L19121.03
also known as L19121-03
Price (USD)
54.65
Special offer
Online exclusive
Ends: 30-Jun-2026
64.70
Save 10.05 (16%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
54.65
Special offer
Online exclusive
Ends: 30-Jun-2026
64.70
Save 10.05 (16%)
Each
Chemical Identifiers
CAS66399-30-2
IUPAC Name1-(4-fluorophenyl)ethan-1-amine
Molecular FormulaC8H10FN
InChI KeyQGCLEUGNYRXBMZ-UHFFFAOYNA-N
SMILESCC(N)C1=CC=C(F)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Optical Rotation-33.0? ± 1.0? (neat)
Enantiomeric excess≥98.5%
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
View more
(S)-(-)-1-(4-Fluorophenyl)ethylamine is used in organic synthesis. It acts as a ligand and form a coordination complex, diacetato-2-O-bis[(S)-1-(4-fluorophenyl)ethylamine-N]palladium(II) by reacting with palladium(II) acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-1-(4-Fluorophenyl)ethylamine is used in organic synthesis. It acts as a ligand and form a coordination complex, diacetato-2-O-bis[(S)-1-(4-fluorophenyl)ethylamine-N]palladium(II) by reacting with palladium(II) acetate.

Solubility
Miscible with dimethyl sulfoxide.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Li, G.; Ren, J.; Iwaki, H.; Zhang, D.; Hasegawa, Y.; Wu, Q.; Feng, J.; Lau, P. C. K.; Zhu, D. Substrate profiling of cyclohexylamine oxidase and its mutants reveals new biocatalytic potential in deracemization of racemic amines. Appl. Microbiol. Biotechnol. 2014, 98 (4), 1681-1689.
  2. Qin, J.; Dhondi, P.; Huang, X.; Mandal, M.; Zhao, Z.; Pissarnitski, D.; Zhou, W.; Aslanian, R.; Zhu, Z.; Greenlee, W.; Clader, J.; Zhang, L.; Cohen-Williams, M.; Jones, N.; Hyde, L.; Palani, A. Discovery of fused 5,6-bicyclic heterocycles as gamma-secretase modulators. Bioorg. Med. Chem. Lett. 2011, 21 (2), 664-669.