(1R)-(-)-Menthyl glyoxylate monohydrate, 98%
(1R)-(-)-Menthyl glyoxylate monohydrate, 98%
(1R)-(-)-Menthyl glyoxylate monohydrate, 98%
(1R)-(-)-Menthyl glyoxylate monohydrate, 98%
Thermo Scientific Chemicals

(1R)-(-)-Menthyl glyoxylate monohydrate, 98%

CAS: 111969-64-3 | C12H22O4 | 230.304 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number L19208.14
also known as L19208-14
Price (USD)
79.65
Online Exclusive
88.10
Save 8.45 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
79.65
Online Exclusive
88.10
Save 8.45 (10%)
Each
Add to cart
Chemical Identifiers
CAS111969-64-3
IUPAC Name5-methyl-2-(propan-2-yl)cyclohexyl 2,2-dihydroxyacetate
Molecular FormulaC12H22O4
InChI KeyBWZMJRSMHQDFIT-UHFFFAOYNA-N
SMILESCC(C)C1CCC(C)CC1OC(=O)C(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (GC)>97.5%
Optical Rotation-75 ? 3? (c=1 in methanol)
Formpowder
Water Content (Karl Fischer Titration)7.0-8.7%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only