2-Thiopheneacetamide, 98%
2-Thiopheneacetamide, 98%
2-Thiopheneacetamide, 98%
Thermo Scientific Chemicals

2-Thiopheneacetamide, 98%

CAS: 4461-29-4 | C6H7NOS | 141.188 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number L19221.22
also known as L19221-22
Price (USD)
176.65
Special offer
Online exclusive
Ends: 30-Jun-2026
208.00
Save 31.35 (15%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
176.65
Special offer
Online exclusive
Ends: 30-Jun-2026
208.00
Save 31.35 (15%)
Each
Chemical Identifiers
CAS4461-29-4
IUPAC Name2-(thiophen-2-yl)acetamide
Molecular FormulaC6H7NOS
InChI KeyUUPZTFTUZUQRQT-UHFFFAOYSA-N
SMILESNC(=O)CC1=CC=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)144.0-150.0?C
Appearance (Color)White to brown
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only