1-Boc-3-piperidone, 97%
1-Boc-3-piperidone, 97%
1-Boc-3-piperidone, 97%
Thermo Scientific Chemicals

1-Boc-3-piperidone, 97%

CAS: 98977-36-7 | C10H17NO3 | 199.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L19276.03
also known as L19276-03
Price (USD)
20.65
Special offer
Online exclusive
Ends: 30-Jun-2026
23.60
Save 2.95 (13%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
20.65
Special offer
Online exclusive
Ends: 30-Jun-2026
23.60
Save 2.95 (13%)
Each
Chemical Identifiers
CAS98977-36-7
IUPAC Nametert-butyl 3-oxopiperidine-1-carboxylate
Molecular FormulaC10H17NO3
InChI KeyRIFXIGDBUBXKEI-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)N1CCCC(=O)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow
FormCrystals or powder or crystalline powder or lumps or fused solid
Assay (GC)≥96.0%
1-Boc-3-piperidone, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-3-piperidone, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
It is insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

General References:

  1. RM Angell.; FL Atkinson.; MJ Brown.; TT Chuang. N-(3-Cyano-4, 5, 6, 7-tetrahydro-1-benzothien-2-yl) amides as potent, selective, inhibitors of JNK2 and JNK3. Bioorganic & Medicinal Chemistry Letters. 200717 (5), 1296-1301.
  2. DC Cole.; WJ Lennox.; JR Stock.; JW Ellingboe. Conformationally constrained N 1-arylsulfonyltryptamine derivatives as 5-HT 6 receptor antagonists. Bioorganic & Medicinal Chemistry Letters. 200515 (21), 1296-1301.