5-Aminomethyl-2-chloropyridine, 97%
5-Aminomethyl-2-chloropyridine, 97%
5-Aminomethyl-2-chloropyridine, 97%
Thermo Scientific Chemicals

5-Aminomethyl-2-chloropyridine, 97%

CAS: 97004-04-1 | C6H7ClN2 | 142.586 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L19283.14
also known as L19283-14
Price (USD)
572.65
Special offer
Online exclusive
Ends: 30-Jun-2026
674.00
Save 101.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
572.65
Special offer
Online exclusive
Ends: 30-Jun-2026
674.00
Save 101.35 (15%)
Each
Chemical Identifiers
CAS97004-04-1
IUPAC Name1-(6-chloropyridin-3-yl)methanamine
Molecular FormulaC6H7ClN2
InChI KeyXPARFBOWIYMLMY-UHFFFAOYSA-N
SMILESNCC1=CN=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point25-33?C (clear to cloudy melt)
FormPowder or Fused solid
Appearance (Color)White to yellow or pale cream to pale brown
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only