(S)-(+)-2-Aminononane, ChiPros 99+%, ee 99+%
Thermo Scientific Chemicals

(S)-(+)-2-Aminononane, ChiPros 99+%, ee 99+%

CAS: 869278-88-6 | C9H21N | 143.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number L19311.03
also known as L19311-03
Price (USD)
49.65
Online Exclusive
54.50
Save 4.85 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
49.65
Online Exclusive
54.50
Save 4.85 (9%)
Each
Chemical Identifiers
CAS869278-88-6
SpecificationsSpecification SheetSpecification Sheet
Enantiomeric excess>99.0%
Appearance (Color)Clear, colorless
FormLiquid
Assay (GC)>99.0%
Optical Rotation+6 ? 0.5? (neat)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only