2-Fluorobenzothiazole, 99%
2-Fluorobenzothiazole, 99%
2-Fluorobenzothiazole, 99%
Thermo Scientific Chemicals

2-Fluorobenzothiazole, 99%

CAS: 1123-98-4 | C7H4FNS | 153.174 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number L19316.03
also known as L19316-03
Price (USD)
100.65
Online Exclusive
112.00
Save 11.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
100.65
Online Exclusive
112.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS1123-98-4
IUPAC Name2-fluoro-1,3-benzothiazole
Molecular FormulaC7H4FNS
InChI KeyQVWCHVAUHZEAAT-UHFFFAOYSA-N
SMILESFC1=NC2=CC=CC=C2S1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.5%
Refractive Index1.5825-1.5855 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only