3-Chloro-4-fluoro-5-nitrobenzotrifluoride, 98%, Thermo Scientific Chemicals
3-Chloro-4-fluoro-5-nitrobenzotrifluoride, 98%, Thermo Scientific Chemicals
3-Chloro-4-fluoro-5-nitrobenzotrifluoride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Chloro-4-fluoro-5-nitrobenzotrifluoride, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
L19322.06
also known as L19322-06
5 g
Catalog number L19322.06
also known as L19322-06
Price (USD)
108.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
108.00
Each
Chemical Identifiers
CAS101646-02-0
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4800-1.4840 @ 20?C
Appearance (Color)Clear colorless to yellow
Assay (GC)≥97.5%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only