Benzo[b]thiophene-2-carboxaldehyde, 97%
Benzo[b]thiophene-2-carboxaldehyde, 97%
Benzo[b]thiophene-2-carboxaldehyde, 97%
Thermo Scientific Chemicals

Benzo[b]thiophene-2-carboxaldehyde, 97%

CAS: 3541-37-5 | C9H6OS | 162.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L19345.03
also known as L19345-03
Price (USD)
59.65
Online exclusive
65.90
Save 6.25
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
59.65
Online exclusive
65.90
Save 6.25
Each
Chemical Identifiers
CAS3541-37-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to cream to yellow to brown
FormFused solid
Assay (GC)≥96.0%
Free acid (titration)≤1.5%
Melting Point (clear melt)28.0-38.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only