1-Allylpiperazine, 98+%
1-Allylpiperazine, 98+%
1-Allylpiperazine, 98+%
Thermo Scientific Chemicals

1-Allylpiperazine, 98+%

CAS: 13961-36-9 | C7H14N2 | 126.203 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L19391.06
also known as L19391-06
Price (USD)
75.65
Special offer
Online exclusive
Ends: 30-Jun-2026
88.80
Save 13.15 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
75.65
Special offer
Online exclusive
Ends: 30-Jun-2026
88.80
Save 13.15 (15%)
Each
Chemical Identifiers
CAS13961-36-9
IUPAC Name1-(prop-2-en-1-yl)piperazine
Molecular FormulaC7H14N2
InChI KeyZWAQJGHGPPDZSF-UHFFFAOYSA-N
SMILESC=CCN1CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
Appearance (Color)Clear colorless to pale yellow
Refractive Index1.4810-1.4850 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only