(R)-(-)-2-Amino-1-phenylethanol, 97%, ee 98%
(R)-(-)-2-Amino-1-phenylethanol, 97%, ee 98%
(R)-(-)-2-Amino-1-phenylethanol, 97%, ee 98%
Thermo Scientific Chemicals

(R)-(-)-2-Amino-1-phenylethanol, 97%, ee 98%

CAS: 2549-14-6 | C8H11NO | 137.182 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L19403.03
also known as L19403-03
Price (USD)
69.65
Online Exclusive
77.10
Save 7.45 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
69.65
Online Exclusive
77.10
Save 7.45 (10%)
Each
Chemical Identifiers
CAS2549-14-6
IUPAC Name2-amino-1-phenylethan-1-ol
Molecular FormulaC8H11NO
InChI KeyULSIYEODSMZIPX-UHFFFAOYNA-N
SMILESNCC(O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)< 2.0%
Enantiomeric excess> 97.5%
Formpowder
Optical Rotation-43.3 ? 1.5? (c=2, ethanol)
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only