2,3-Diamino-5-bromopyridine, 97%
2,3-Diamino-5-bromopyridine, 97%
2,3-Diamino-5-bromopyridine, 97%
2,3-Diamino-5-bromopyridine, 97%
Thermo Scientific Chemicals

2,3-Diamino-5-bromopyridine, 97%

CAS: 38875-53-5 | C5H7BrN3 | 189.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L19435.14
also known as L19435-14
Price (USD)
54.65
Online Exclusive
60.70
Save 6.05 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
54.65
Online Exclusive
60.70
Save 6.05 (10%)
Each
Add to cart
Chemical Identifiers
CAS38875-53-5
IUPAC Name2,3-diamino-5-bromopyridin-1-ium
Molecular FormulaC5H7BrN3
InChI KeyYRGMYJUKFJPNPD-UHFFFAOYSA-O
SMILESNC1=CC(Br)=C[NH+]=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or needles
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0%
Appearance (Color)Pale yellow to yellow to pale brown to brown to purple
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only