1,4-Benzenedithiol, 97%
1,4-Benzenedithiol, 97%
1,4-Benzenedithiol, 97%
Thermo Scientific Chemicals

1,4-Benzenedithiol, 97%

CAS: 624-39-5 | C6H6S2 | 142.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L19440.MD
also known as L19440-MD
Price (USD)
50.65
Special offer
Online exclusive
Ends: 30-Jun-2026
59.60
Save 8.95 (15%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
50.65
Special offer
Online exclusive
Ends: 30-Jun-2026
59.60
Save 8.95 (15%)
Each
Chemical Identifiers
CAS624-39-5
IUPAC Namebenzene-1,4-dithiol
Molecular FormulaC6H6S2
InChI KeyWYLQRHZSKIDFEP-UHFFFAOYSA-N
SMILESSC1=CC=C(S)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)93.0-102.0?C
1,4-Benzenedithiol is used in single molecule junctions in electronic devices. It is also used to treat silver nano particles and in the preparation of gold electrodes. It finds an application in nanomaterial synthesis and in self-assembled monolayer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,4-Benzenedithiol is used in single molecule junctions in electronic devices. It is also used to treat silver nano particles and in the preparation of gold electrodes. It finds an application in nanomaterial synthesis and in self-assembled monolayer.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

General References:

  1. Matsuhita, R.; Horikawa, M.; Naitoh, Y.; Nakamura, H.; Kiguchi, M. Conductance and SERS Measurement of Benzenedithiol Molecules Bridging Between Au Electrodes. J. Phys. Chem., C 2013,117 (4), 1791-1795.
  2. Vidal-Iglesias, F. J.; Solla-Gullón, J.; Orts, J. M.; Rodes, A.; Pérez, J. M. Spectroelectrochemical Study of the Photoinduced Catalytic Formation of 4,4'-Dimercaptoazobenzene from 4-Aminobenzenethiol Adsorbed on Nanostructured Copper. J. Phys. Chem. C 2015, 119 (22), 12312-12324.