2,6-Bis(trifluoromethyl)pyridine, 97%
2,6-Bis(trifluoromethyl)pyridine, 97%
2,6-Bis(trifluoromethyl)pyridine, 97%
2,6-Bis(trifluoromethyl)pyridine, 97%
Thermo Scientific Chemicals

2,6-Bis(trifluoromethyl)pyridine, 97%

CAS: 455-00-5 | C7H3F6N | 215.098 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L19446.03
also known as L19446-03
Price (USD)
71.65
Special offer
Online exclusive
Ends: 30-Jun-2026
84.40
Save 12.75 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
71.65
Special offer
Online exclusive
Ends: 30-Jun-2026
84.40
Save 12.75 (15%)
Each
Add to cart
Chemical Identifiers
CAS455-00-5
IUPAC Name2,6-bis(trifluoromethyl)pyridine
Molecular FormulaC7H3F6N
InChI KeyYPDVFTXBESQIPJ-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=CC(=N1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)53.5-59.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only