2-Cyano-3-methylpyridine, 98%
2-Cyano-3-methylpyridine, 98%
2-Cyano-3-methylpyridine, 98%
Thermo Scientific Chemicals

2-Cyano-3-methylpyridine, 98%

CAS: 20970-75-6 | C7H6N2 | 118.139 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number L19456.14
also known as L19456-14
Price (USD)
119.65
Online Exclusive
133.00
Save 13.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
119.65
Online Exclusive
133.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS20970-75-6
IUPAC Name3-methylpyridine-2-carbonitrile
Molecular FormulaC7H6N2
InChI KeyWBXZCDIZXWDPBL-UHFFFAOYSA-N
SMILESCC1=CC=CN=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)83.0-89.0?C
Appearance (Color)White to pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only