2,3,5,6-Tetrachloropyridine, 98%
2,3,5,6-Tetrachloropyridine, 98%
2,3,5,6-Tetrachloropyridine, 98%
Thermo Scientific Chemicals

2,3,5,6-Tetrachloropyridine, 98%

CAS: 2402-79-1 | C5HCl4N | 216.87 g/mol
Have Questions?
Catalog NumberQuantity
L19458.03
also known as L19458-03
1 g
Catalog number L19458.03
also known as L19458-03
Price (USD)
14.70
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
14.70
Each
Chemical Identifiers
CAS2402-79-1
IUPAC Name2,3,5,6-tetrachloropyridine
Molecular FormulaC5HCl4N
InChI KeyFATBKZJZAHWCSL-UHFFFAOYSA-N
SMILESClC1=CC(Cl)=C(Cl)N=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)89-94?C
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only