N-Boc-1,3-diaminopropane hydrochloride, 98%, Thermo Scientific Chemicals
N-Boc-1,3-diaminopropane hydrochloride, 98%, Thermo Scientific Chemicals
N-Boc-1,3-diaminopropane hydrochloride, 98%, Thermo Scientific Chemicals
N-Boc-1,3-diaminopropane hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-1,3-diaminopropane hydrochloride, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
L19486.03
also known as L19486-03
1 g
Catalog number L19486.03
also known as L19486-03
Price (USD)
100.65
Online Exclusive
112.00
Save 11.35 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
100.65
Online Exclusive
112.00
Save 11.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS127346-48-9
IUPAC Nametert-butyl N-(3-aminopropyl)carbamate hydrochloride
Molecular FormulaC8H19ClN2O2
InChI KeyWUXOJNUZYOFBMI-UHFFFAOYSA-N
SMILESCl.CC(C)(C)OC(=O)NCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Water Content (Karl Fischer Titration)≤1%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only