(R)-(-)-2-(Methoxymethyl)pyrrolidine, 98+%
(R)-(-)-2-(Methoxymethyl)pyrrolidine, 98+%
(R)-(-)-2-(Methoxymethyl)pyrrolidine, 98+%
Thermo Scientific Chemicals

(R)-(-)-2-(Methoxymethyl)pyrrolidine, 98+%

CAS: 84025-81-0 | C6H13NO | 115.176 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number L19492.06
also known as L19492-06
Price (USD)
398.65
Online Exclusive
443.00
Save 44.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
398.65
Online Exclusive
443.00
Save 44.35 (10%)
Each
Chemical Identifiers
CAS84025-81-0
IUPAC Name2-(methoxymethyl)pyrrolidine
Molecular FormulaC6H13NO
InChI KeyCHPRFKYDQRKRRK-UHFFFAOYNA-N
SMILESCOCC1CCCN1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥98.0%
Optical Rotation-10.5 ± 9.5? (C=2 in benzene)
Refractive Index1.4440-1.4480 @ 20?C
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only