(S)-(-)-3-Hydroxypyrrolidine, 97+%, ee 98+%
(S)-(-)-3-Hydroxypyrrolidine, 97+%, ee 98+%
(S)-(-)-3-Hydroxypyrrolidine, 97+%, ee 98+%
Thermo Scientific Chemicals

(S)-(-)-3-Hydroxypyrrolidine, 97+%, ee 98+%

CAS: 100243-39-8 | C4H10NO | 88.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L19498.03
also known as L19498-03
Price (USD)
161.65
Online Exclusive
179.00
Save 17.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
161.65
Online Exclusive
179.00
Save 17.35 (10%)
Each
Chemical Identifiers
CAS100243-39-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white to yellow
FormCrystals or powder or crystalline powder or lumps or fused solid or clear liquid as melt
Assay (GC)≥97.0%
(S)-(-)-3-Hydroxypyrrolidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-3-Hydroxypyrrolidine is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Fully miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents. It is hygroscopic and sensitive to air.
RUO – Research Use Only

General References:

  1. Zhi Li, Hans-Jürgen Feiten, Jan B van Beilen, Wouter Duetz, Bernard Witholtt. Preparation of optically active N-benzyl-3-hydroxypyrrolidine by enzymatic hydroxylation. Tetrahedron: Asymmetry. 1999, 10 (7), 1323-1333.
  2. Zhi Li.; Hans-Jürgen Feiten.; Dongliang Chang.; Wouter A. Duetz.; Jan B. van Beilen.; Bernard Witholt. Preparation of (R)- and (S)-N-Protected 3-Hydroxypyrrolidines by Hydroxylation with Sphingomonas sp. HXN-200, a Highly Active, Regio- and Stereoselective, and Easy to Handle Biocatalyst. J. Org. Chem. 2001, 66 (25), 8424-8430.
  3. Product of Toray Industries, Inc.