4-(Methylsulfonyl)phenylacetic acid, 97%
4-(Methylsulfonyl)phenylacetic acid, 97%
4-(Methylsulfonyl)phenylacetic acid, 97%
4-(Methylsulfonyl)phenylacetic acid, 97%
Thermo Scientific Chemicals

4-(Methylsulfonyl)phenylacetic acid, 97%

CAS: 90536-66-6 | C9H10O4S | 214.235 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L19504.14
also known as L19504-14
Price (USD)
170.65
Online Exclusive
189.00
Save 18.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
170.65
Online Exclusive
189.00
Save 18.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS90536-66-6
IUPAC Name2-(4-methanesulfonylphenyl)acetate
Molecular FormulaC9H9O4S
InChI KeyHGGWOSYNRVOQJH-UHFFFAOYSA-M
SMILESCS(=O)(=O)C1=CC=C(CC([O-])=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to yellow
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (HPLC)≥96.0%
Melting Point (clear melt)130.0-140.0?C
4-(Methylsulfonyl)phenylacetic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Methylsulfonyl)phenylacetic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
RUO – Research Use Only
Brenda Pipik.; Guo-Jie Ho.; J. Michael Williams.; David A. A Preferred Synthesis of 1,2,4-Oxadiazoles. Synthetic Communications: An International Journal for Rapid Communication of Synthetic Organic Chemistry. 2004, 34 (10), 1863-1870.