1,3-Difluoro-5-iodobenzene, 98%
1,3-Difluoro-5-iodobenzene, 98%
1,3-Difluoro-5-iodobenzene, 98%
1,3-Difluoro-5-iodobenzene, 98%
Thermo Scientific Chemicals

1,3-Difluoro-5-iodobenzene, 98%

CAS: 2265-91-0 | C6H3F2I | 239.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L19510.03
also known as L19510-03
Price (USD)
36.65
Online Exclusive
41.00
Save 4.35 (11%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
36.65
Online Exclusive
41.00
Save 4.35 (11%)
Each
Add to cart
Chemical Identifiers
CAS2265-91-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥98.0%
Identification (FTIR)Conforms
Refractive Index1.5520-1.5560 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only