2-Indanylboronic acid pinacol ester, 95%
2-Indanylboronic acid pinacol ester, 95%
2-Indanylboronic acid pinacol ester, 95%
Thermo Scientific Chemicals

2-Indanylboronic acid pinacol ester, 95%

CAS: 608534-44-7 | C15H21BO2 | 244.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L19535.03
also known as L19535-03
Price (USD)
107.65
Special offer
Online exclusive
Ends: 30-Jun-2026
126.00
Save 18.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
107.65
Special offer
Online exclusive
Ends: 30-Jun-2026
126.00
Save 18.35 (15%)
Each
Chemical Identifiers
CAS608534-44-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Refractive Index1.5010-1.5070 @ 20?C
Assay (GC)≥94.0%
Boronic acids react with alcohols, with loss of water, to form boronic esters. With simple alcohols, the products are very susceptible to hydrolysis, but with 1,2- and 1,3-diols, the resulting cyclic boronates (1,3,2- dioxaborolanes and 1,3,2-dioxaborinanes) are stable enough to be isolated.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Boronic acids react with alcohols, with loss of water, to form boronic esters. With simple alcohols, the products are very susceptible to hydrolysis, but with 1,2- and 1,3-diols, the resulting cyclic boronates (1,3,2- dioxaborolanes and 1,3,2-dioxaborinanes) are stable enough to be isolated.

Solubility
Difficult to mix in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.
RUO – Research Use Only