3,5-Difluoropyridine, 98%
3,5-Difluoropyridine, 98%
3,5-Difluoropyridine, 98%
3,5-Difluoropyridine, 98%
Thermo Scientific Chemicals

3,5-Difluoropyridine, 98%

CAS: 71902-33-5 | C5H3F2N | 115.083 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number L19539.06
Price (USD)
170.00
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
170.00
Each
Add to cart
Specifications
Beilstein7914363
CAS71902-33-5
Chemical Name or Material3,5-Difluoropyridine
DOT InformationTransport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Density1.268
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only