3-Chloro-2,5,6-trifluoropyridine, 98+%
3-Chloro-2,5,6-trifluoropyridine, 98+%
3-Chloro-2,5,6-trifluoropyridine, 98+%
3-Chloro-2,5,6-trifluoropyridine, 98+%
Thermo Scientific Chemicals

3-Chloro-2,5,6-trifluoropyridine, 98+%

CAS: 2879-42-7 | C5HClF3N | 167.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number L19543.06
also known as L19543-06
Price (USD)
82.65
Online Exclusive
92.20
Save 9.55 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
82.65
Online Exclusive
92.20
Save 9.55 (10%)
Each
Add to cart
Chemical Identifiers
CAS2879-42-7
IUPAC Name3-chloro-2,5,6-trifluoropyridine
Molecular FormulaC5HClF3N
InChI KeyOROKHXKJSDPRET-UHFFFAOYSA-N
SMILESFC1=CC(Cl)=C(F)N=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.0%
Refractive Index1.4545-1.4585 @ 20?C
FormLiquid
3-Chloro-2,5,6-trifluoropyridine is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloro-2,5,6-trifluoropyridine is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. R.E. Banks.; A. Prakash.; N.D. Venayak. Studies in azide chemistry. Part IX [1]. Investigations involving fluorinated azidopyrimidines and 4-azido-3-chloro-2,5,6-trifluoropyridine. Journal of Fluorine Chemistry. 1980, 16 (4), 325-338.
  2. Carla Bobbio.; Thierry Rausis.; Manfred Schlosser Prof. Removal of Fluorine from and Introduction of Fluorine into Polyhalopyridines: An Exercise in Nucleophilic Hetarenic Substitution. Chemistry-A European Journal. 2005, 11 (6), 1903-1910.