4-Chloro-3-(trifluoromethyl)pyridine hydrochloride, 97%
4-Chloro-3-(trifluoromethyl)pyridine hydrochloride, 97%
4-Chloro-3-(trifluoromethyl)pyridine hydrochloride, 97%
Thermo Scientific Chemicals

4-Chloro-3-(trifluoromethyl)pyridine hydrochloride, 97%

CAS: 732306-24-0 | C6H4Cl2F3N | 218.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number L19556.03
also known as L19556-03
Price (USD)
198.65
Special offer
Online exclusive
Ends: 30-Jun-2026
233.00
Save 34.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
198.65
Special offer
Online exclusive
Ends: 30-Jun-2026
233.00
Save 34.35 (15%)
Each
Chemical Identifiers
CAS732306-24-0
IUPAC Name4-chloro-3-(trifluoromethyl)pyridine hydrochloride
Molecular FormulaC6H4Cl2F3N
InChI KeyHCKXJUZITVCDND-UHFFFAOYSA-N
SMILESCl.FC(F)(F)C1=C(Cl)C=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or lumps
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only